(R)-(-)-2,2'-Bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%

Supplier: Ambeed
Warning

A127511-100MG A127511-1G A127511-50MG
77246-370EA 132.87 USD
77246-370 77246-372 77246-374
(R)-(-)-2,2'-Bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl 98%
(R)-(-)-2,2'-Bis[di(3,5-di-t-butyl-4-methoxyphenyl)phosphino]-6,6'-dimethoxy-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₇₄H₁₀₄O₆P₂
MW: 1151.5615 g/mol
MDL Number: MFCD09265029
CAS Number: 352655-61-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR