Methyl(triphenylphosphine)gold(I) 98%

Supplier: Ambeed
Warning

A127339-1G A127339-250MG
77246-360EA 368.74 USD
77246-360 77246-362
Methyl(triphenylphosphine)gold(I) 98%
Methyl(triphenylphosphine)gold(I)

Precaución: for Research Use Only

Formula: C₁₉H₁₉AuP
MW: 475.2945 g/mol
MDL Number: MFCD07369042
CAS Number: 23108-72-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR