4-(3,4-Dichlorophenyl)thiazol-2-amine 97%
Supplier: Ambeed
|
Warning
|
A127328-10G
A127328-1G
A127328-25G
A127328-5G
77285-156EA
95
USD
77285-156
77285-158
77285-160
77285-162
4-(3,4-Dichlorophenyl)thiazol-2-amine 97%
4-(3,4-Dichlorophenyl)thiazol-2-amine
Precaución: for Research Use Only
Formula:
C₉H₆Cl₂N₂S |
MDL Number:
MFCD00466323 CAS Number: 39893-80-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...