4,5-Dichloro-2-phenyl-3(2H)-pyridazinone 97%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dichloro-2-phenylpyridazin-3(2H)-one

A127156-1G A127156-250MG A127156-25G A127156-5G
77234-354EA 12.36 USD
77234-354 77234-356 77234-358 77234-360
4,5-Dichloro-2-phenyl-3(2H)-pyridazinone 97%
4,5-Dichloro-2-phenyl-3(2H)-pyridazinone

Precaución: for Research Use Only

Formula: C₁₀H₆Cl₂N₂O
MW: 241.07 g/mol
MDL Number: MFCD00006470
CAS Number: 1698-53-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR