1-Ethyl-2,3-piperazinedione 97%

Supplier: Ambeed
Warning

Synonyms: 1-Ethylpiperazine-2,3-dione

A127119-10G A127119-5G
77285-144EA 19.57 USD
77285-144 77285-146
1-Ethyl-2,3-piperazinedione 97%
1-Ethyl-2,3-piperazinedione

Precaución: for Research Use Only

Formula: C₆H₁₀N₂O₂
MW: 142.16 g/mol
MDL Number: MFCD00051825
CAS Number: 59702-31-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR