1-Methoxy-4-(trans-4-propylcyclohexyl)benzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Methoxy-4-(trans-4-N-propylcyclohexyl)benzene
A127114-10G
A127114-25G
A127114-1G
77271-830EA
13
USD
77271-830
77271-832
77747-910
1-Methoxy-4-(trans-4-propylcyclohexyl)benzene 98%
1-Methoxy-4-(trans-4-propylcyclohexyl)benzene
Precaución: for Research Use Only
Formula:
C₁₆H₂₄O MW: 232.37 g/mol |
MDL Number:
MFCD06658193 CAS Number: 81936-32-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...