2-Bromo-6-(methoxycarbonyl)benzoic acid 97%

Supplier: Ambeed
Warning

A126823-100MG A126823-250MG
77246-352EA 58 USD
77246-352 77246-354
2-Bromo-6-(methoxycarbonyl)benzoic acid 97%
2-Bromo-6-(methoxycarbonyl)benzoic acid

Precaución: for Research Use Only

Formula: C₉H₇BrO₄
MDL Number: MFCD22490791
CAS Number: 912457-18-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR