4-(2-Oxo-2,3-dihydro-1H-1,3-benzimidazol-1-yl)butanoic acid 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-(2-Oxo-2,3-dihydro-1H-benzo[d]imidazol-1-yl)butanoic acid
A126619-100G
A126619-1G
A126619-250MG
A126619-25G
A126619-5G
77234-344EA
65
USD
77234-344
77234-346
77234-348
77234-350
77234-352
4-(2-Oxo-2,3-dihydro-1H-1,3-benzimidazol-1-yl)butanoic acid 98%
4-(2-Oxo-2,3-dihydro-1H-1,3-benzimidazol-1-yl)butanoic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₂N₂O₃ |
MDL Number:
MFCD06656423 CAS Number: 3273-68-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...