BTZ043 98+%

Supplier: Ambeed
Warning

Synonyms: (S)-2-(2-methyl-1,4-dioxa-8-azaspiro[4.5]decan-8-yl)-8-nitro-6-(trifluoromethyl)-4H-benzo[e][1,3]thiazin-4-one

A126564-100MG A126564-10MG A126564-50MG A126564-5MG
77234-328EA 94.76 USD
77234-328 77234-330 77234-332 77234-334
BTZ043 98+%
BTZ043

Precaución: for Research Use Only

Formula: C₁₇H₁₆F₃N₃O₅S
MW: 431.39 g/mol
MDL Number: MFCD17215196
CAS Number: 1161233-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR