16-Bromo-1-hexadecanol 95%

Supplier: Ambeed
Warning

Synonyms: 16-Bromohexadecan-1-ol

A126535-1G A126535-250MG A126535-5G A126535-100MG
77271-810EA 161.71 USD
77271-810 77271-812 77571-582 77747-904
16-Bromo-1-hexadecanol 95%
16-Bromo-1-hexadecanol

Precaución: for Research Use Only

Formula: HO(CH₂)₁₅CH₂Br
MW: 321.34 g/mol
MDL Number: MFCD01076224
CAS Number: 59101-28-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR