Methyl 3-oxocyclohexanecarboxylate 95%

Supplier: Ambeed
Warning

A126461-100MG A126461-1G A126461-250MG A126461-5G A126461-25G
77285-128EA 9.27 USD
77285-128 77285-130 77285-132 77285-134 77747-903
Methyl 3-oxocyclohexanecarboxylate 95%
Methyl 3-oxocyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₂O₃
MW: 156.179 g/mol
MDL Number: MFCD09031996
CAS Number: 13148-83-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR