Ethyl 3-Oxo-3-(2-thienyl)propionate 97%

Supplier: Ambeed
Warning

A126368-10G A126368-1G A126368-250MG A126368-25G A126368-5G
77246-336EA 37 USD
77246-336 77246-338 77246-340 77246-342 77246-344
Ethyl 3-Oxo-3-(2-thienyl)propionate 97%
Ethyl 3-Oxo-3-(2-thienyl)propionate

Precaución: for Research Use Only

Formula: C₉H₁₀O₃S
MDL Number: MFCD00542649
CAS Number: 13669-10-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR