S63845 98%

Supplier: Ambeed
Warning

A126292-10MG A126292-1MG A126292-25MG A126292-5MG
77258-290EA 220.01 USD
77258-290 77258-292 77258-294 77258-296
S63845 98%
S63845

Precaución: for Research Use Only

Formula: C₃₉H₃₇ClF₄N₆O₆S
MDL Number: MFCD31382374
CAS Number: 1799633-27-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR