4-Cyano-2,6-difluorobenzoic acid 97%

Supplier: Ambeed
Warning

A126285-1G A126285-250MG
77285-120EA 180 USD
77285-120 77285-122
4-Cyano-2,6-difluorobenzoic acid 97%
4-Cyano-2,6-difluorobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₃F₂NO₂
MDL Number: MFCD09835199
CAS Number: 181073-82-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR