ω-Bromo-3-hydroxyacetophenone 96%

Supplier: Ambeed
Danger

A126120-100MG A126120-1G A126120-250MG A126120-25G A126120-5G
77258-278EA 5 USD
77258-278 77258-280 77258-282 77258-284 77571-552
ω-Bromo-3-hydroxyacetophenone 96%
ω-Bromo-3-hydroxyacetophenone

Precaución: for Research Use Only

Formula: C₈H₇BrO₂
MDL Number: MFCD05664326
CAS Number: 2491-37-4
UN: 1759
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR