5-(2-Hydroxyethyl)amino-o-cresol 97%

Supplier: Ambeed
Warning

Synonyms: 5-((2-Hydroxyethyl)amino)-2-methylphenol, 5-(2-Hydroxyethyl)amino-2-methylphenol

A125889-10G A125889-1G A125889-5G
77271-778EA 21.63 USD
77271-778 77271-780 77271-782
5-(2-Hydroxyethyl)amino-o-cresol 97%
5-(2-Hydroxyethyl)amino-o-cresol

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
MDL Number: MFCD01707346
CAS Number: 55302-96-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR