Pivalic acid hydrazide 95%

Supplier: Ambeed
Danger

Synonyms: Pivalohydrazide, Pivaloyl hydrazide

A125747-1G A125747-5G
77246-328EA 14 USD
77246-328 77566-430
Pivalic acid hydrazide 95%
Pivalic acid hydrazide

Precaución: For research use only.

Formula: C₅H₁₂N₂O
MDL Number: MFCD01310839
CAS Number: 42826-42-6
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR