2-Formyl-4-methoxyphenylboronic acid 97%

Supplier: Ambeed
Warning

A125682-10G A125682-1G A125682-250MG A125682-5G
77246-322EA 99 USD
77246-322 77246-324 77246-326 77571-532
2-Formyl-4-methoxyphenylboronic acid 97%
2-Formyl-4-methoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₉BO₄
MDL Number: MFCD01319043
CAS Number: 139962-95-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR