6-Amino-2-(methylthio)pyrimidin-4-ol 98%

Supplier: Ambeed
Warning

Synonyms: 4-Amino-2-(methylthio)-6-pyrimidinol

A125519-1G A125519-250MG A125519-25G
77285-094EA 14.42 USD
77285-094 77285-096 77285-098
6-Amino-2-(methylthio)pyrimidin-4-ol 98%
6-Amino-2-(methylthio)pyrimidin-4-ol

Precaución: for Research Use Only

Formula: C₅H₇N₃OS
MW: 157.194 g/mol
MDL Number: MFCD00023240
CAS Number: 1074-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR