4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta[4,5]thieno[2,3-d]pyrimidine 97%

Supplier: Ambeed
Warning

A125516-100MG A125516-250MG
77285-090EA 22.66 USD
77285-090 77285-092
4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta[4,5]thieno[2,3-d]pyrimidine 97%
4-Chloro-6,7,8,9-tetrahydro-5H-cyclohepta[4,5]thieno[2,3-d]pyrimidine

Precaución: for Research Use Only

Formula: C₁₁H₁₁ClN₂S
MDL Number: MFCD05865130
CAS Number: 40106-58-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR