(2R)-1,4-Dioxane-2-methanol 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
(R)-(1,4-Dioxan-2-yl)methanol
A125505-100MG
A125505-10G
A125505-1G
A125505-250MG
A125505-25G
A125505-5G
77271-748EA
15.45
USD
77271-748
77271-750
77271-752
77271-754
77271-756
77271-758
(2R)-1,4-Dioxane-2-methanol 97%
(2R)-1,4-Dioxane-2-methanol
Precaución: for Research Use Only
Formula:
C₅H₁₀O₃ MW: 118.131 g/mol |
MDL Number:
MFCD16620507 CAS Number: 406913-88-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...