7-Bromo-1-methyl-2-phenyl-1H-imidazo[4,5-c]pyridine 97%

Supplier: Ambeed
Warning

A125463-250MG A125463-50MG
77246-306EA 241.02 USD
77246-306 77246-308
7-Bromo-1-methyl-2-phenyl-1H-imidazo[4,5-c]pyridine 97%
7-Bromo-1-methyl-2-phenyl-1H-imidazo[4,5-c]pyridine

Precaución: for Research Use Only

Formula: C₁₃H₁₀BrN₃
MW: 288.14 g/mol
MDL Number: MFCD11046884
CAS Number: 1044772-00-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR