rel-3-Chloro-N-((1r,4r)-4-(methylamino)cyclohexyl)-N-(3-(pyridin-4-yl)benzyl)benzo[b]thiophene-2-carboxamide hydrochloride 98+%
Supplier: Ambeed
|
Warning
|
Synonyms:
SAG HCl
A125368-10MG
A125368-1MG
A125368-25MG
A125368-5MG
A125368-100MG
A125368-50MG
77258-258EA
40
USD
77258-258
77258-260
77258-262
77258-264
77747-899
77747-900
rel-3-Chloro-N-((1r,4r)-4-(methylamino)cyclohexyl)-N-(3-(pyridin-4-yl)benzyl)benzo[b]thiophene-2-carboxamide hydrochloride 98+%
rel-3-Chloro-N-((1r,4r)-4-(methylamino)cyclohexyl)-N-(3-(pyridin-4-yl)benzyl)benzo[b]thiophene-2-carboxamide hydrochloride
Precaución: for Research Use Only
Formula:
C₂₈H₂₉Cl₂N₃OS |
CAS Number:
2095432-58-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...