4,7-Bis(5-trimethylstannyl-2-thienyl)-2,1,3-benzothiadiazole 97%

Supplier: Ambeed
Danger

Synonyms: 4,7-Bis(5-(trimethylstannyl)thiophen-2-yl)benzo[c][1,2,5]thiadiazole

A125161-1G A125161-250MG
77234-302EA 237.93 USD
77234-302 77234-304
4,7-Bis(5-trimethylstannyl-2-thienyl)-2,1,3-benzothiadiazole 97%
4,7-Bis(5-trimethylstannyl-2-thienyl)-2,1,3-benzothiadiazole

Precaución: for Research Use Only

Formula: C₂₀H₂₄N₂S₃Sn₂
MW: 626.03 g/mol
MDL Number: MFCD28049209
CAS Number: 1025451-57-3
UN: 3146
ADR: 6.1,Ⅰ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR