4-Ethoxybenzyl alcohol 98%

Supplier: Ambeed
Warning

A125154-1G A125154-25G A125154-5G
77285-076EA 15 USD
77285-076 77285-078 77285-080
4-Ethoxybenzyl alcohol 98%
4-Ethoxybenzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₁₂O₂
MDL Number: MFCD00004655
CAS Number: 6214-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR