4,4-Pentamethylene-2-pyrrolidone 95%

Supplier: Ambeed
Warning

A125052-100G A125052-10G A125052-25G A125052-500G A125052-5G
77246-286EA 58 USD
77246-286 77246-288 77246-290 77246-292 77246-294
4,4-Pentamethylene-2-pyrrolidone 95%
4,4-Pentamethylene-2-pyrrolidone

Precaución: for Research Use Only

Formula: C₉H₁₅NO
MDL Number: MFCD00177938
CAS Number: 64744-50-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR