Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate 98%

Supplier: Ambeed
Warning

A125049-100G A125049-25G A125049-1000G A125049-500G
77246-282EA 14 USD
77246-282 77246-284 77592-424 77592-426
Dimethyl-1,4-cyclohexanedione-2,5-dicarboxylate 98%
Dimethyl 1,4-cyclohexanedione-2,5-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₆
MDL Number: MFCD00001607
CAS Number: 6289-46-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR