Bafetinib 98+%

Supplier: Ambeed
Warning

Synonyms: (S)-N-(3-([4,5'-Bipyrimidin]-2-ylamino)-4-methylphenyl)-4-((3-(dimethylamino)pyrrolidin-1-yl)methyl)-3-(trifluoromethyl)benzamide, NS-187

A124910-100MG A124910-10MG A124910-1MG A124910-5MG
77258-222EA 397.58 USD
77258-222 77614-980 77614-982 77747-895
Bafetinib 98+%
Bafetinib

Precaución: for Research Use Only

Formula: C₃₀H₃₁F₃N₈O
MW: 576.6 g/mol
MDL Number: MFCD18633200
CAS Number: 859212-16-1
New Product

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR