3-Amino-1,1'-biphenyl-4-carboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-Amino-[1,1'-biphenyl]-4-carboxylic acid

A124862-100MG A124862-1G A124862-5G A124862-250MG
77271-742EA 27 USD
77271-742 77271-744 77271-746 77747-894
3-Amino-1,1'-biphenyl-4-carboxylic acid 97%
3-Amino-1,1'-biphenyl-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁NO₂
MDL Number: MFCD00577380
CAS Number: 4445-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR