1-(4-Aminopyridin-2-yl)ethanone 97%

Supplier: Ambeed
Warning

A124781-100MG A124781-250MG
77234-296EA 78.28 USD
77234-296 77234-298
1-(4-Aminopyridin-2-yl)ethanone 97%
1-(4-Aminopyridin-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₇H₈N₂O
MW: 136.15 g/mol
MDL Number: MFCD18255782
CAS Number: 1256785-86-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR