tert-Butyl 5-aminoisoindoline-2-carboxylate oxalate(2:1) 97%

Supplier: Ambeed
Warning

A124736-1G A124736-5G
77258-216EA 114.33 USD
77258-216 77258-218
tert-Butyl 5-aminoisoindoline-2-carboxylate oxalate(2:1) 97%
tert-Butyl 5-aminoisoindoline-2-carboxylate oxalate(2:1)

Precaución: for Research Use Only

Formula: C₂₈H₃₈N₄O₈
MW: 558.62 g/mol
MDL Number: MFCD30609512
CAS Number: 2061979-87-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR