5-Nitroindole-2-carboxylic acid 97%
Supplier: Ambeed
|
Warning
|
A124735-100G
A124735-10G
A124735-1G
A124735-250MG
A124735-25G
A124735-5G
77271-730EA
935
USD
77271-730
77271-732
77271-734
77271-736
77271-738
77271-740
5-Nitroindole-2-carboxylic acid 97%
5-Nitroindole-2-carboxylic acid
Precaución: for Research Use Only
Formula:
C₉H₆N₂O₄ |
MDL Number:
MFCD00236372 CAS Number: 16730-20-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...