2-(2-(1H-Indol-3-yl)acetamido)pentanedioic acid 95%

Supplier: Ambeed
Warning

Synonyms: N-DL-(3-IndolylAc)-Glu-OH

A124662-100MG A124662-1G A124662-250MG
77258-204EA 14.42 USD
77258-204 77258-206 77258-208
2-(2-(1H-Indol-3-yl)acetamido)pentanedioic acid 95%
2-(2-(1H-Indol-3-yl)acetamido)pentanedioic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₂O₅
MW: 304.30 g/mol
MDL Number: MFCD07637312
CAS Number: 192430-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR