5-Amino-[1,1'-biphenyl]-2-ol 97%

Supplier: Ambeed
Warning

A124638-100MG A124638-1G A124638-250MG
77246-266EA 27 USD
77246-266 77246-268 77246-270
5-Amino-[1,1'-biphenyl]-2-ol 97%

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO
MDL Number: MFCD08361726
CAS Number: 19434-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR