2-Bromo-5-fluoroaniline 98%

Supplier: Ambeed
Warning

A124618-100G A124618-10G A124618-25G A124618-500G A124618-5G
77285-056EA 48 USD
77285-056 77285-058 77285-060 77285-062 77285-064
2-Bromo-5-fluoroaniline 98%
2-Bromo-5-fluoroaniline

Precaución: for Research Use Only

Formula: C₆H₅BrFN
MDL Number: MFCD00070750
CAS Number: 1003-99-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR