2-Amino-4-methoxybenzothiazole 95%

Supplier: Ambeed
Warning

Synonyms: 4-Methoxybenzothiazol-2-ylamine

A124604-100MG A124604-10G A124604-1G A124604-250MG A124604-25G A124604-5G
77246-248EA 5.15 USD
77246-248 77246-250 77246-252 77246-254 77246-256 77246-258
2-Amino-4-methoxybenzothiazole 95%
2-Amino-4-methoxybenzothiazole

Precaución: for Research Use Only

Formula: C₈H₈N₂OS
MW: 180.23 g/mol
MDL Number: MFCD00005792
CAS Number: 5464-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR