2-Bromo-6-(bromomethyl)pyridine 98%
Supplier: Ambeed
|
Danger
|
A124523-100MG
A124523-1G
A124523-250MG
A124523-5G
77258-194EA
15.45
USD
77258-194
77258-196
77258-198
77258-200
2-Bromo-6-(bromomethyl)pyridine 98%
2-Bromo-6-(bromomethyl)pyridine
Precaución: for Research Use Only
Formula:
C₆H₅Br₂N MW: 250.9200 g/mol |
MDL Number:
MFCD07368202 CAS Number: 83004-10-8 UN: 3261 ADR: 8,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...