PF-03394197 98%

Supplier: Ambeed
Warning

Synonyms: Oclacitinib, N-Methyl-1-(trans-4-(methyl(7H-pyrrolo[2,3-d]pyrimidin-4-yl)amino)cyclohexyl)methanesulphonamide

A124514-10MG A124514-2MG A124514-5MG
77234-276EA 21 USD
77234-276 77234-278 77234-280 77234-282
PF-03394197 98%
PF-03394197

Precaución: for Research Use Only

Formula: C₁₅H₂₃N₅O₂S
MW: 337.44 g/mol
MDL Number: MFCD25976611
CAS Number: 1208319-26-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR