5-Amino-1-phenyl-1H-pyrazole 97%

Supplier: Ambeed
Warning

Synonyms: 1-Phenyl-1H-pyrazol-5-amine, 5-Amino-1-phenylpyrazole

A124478-10G A124478-1G A124478-5G
77271-724EA 79.31 USD
77271-724 77271-726 77614-976
5-Amino-1-phenyl-1H-pyrazole 97%
5-Amino-1-phenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₉H₉N₃
MW: 159.1900 g/mol
MDL Number: MFCD00020734
CAS Number: 826-85-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR