(2-Chloro-5-fluorophenyl)acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-5-fluorophenylacetic acid

A124429-10G A124429-1G A124429-25G A124429-5G
77258-184EA 87 USD
77258-184 77258-186 77258-188 77258-190
(2-Chloro-5-fluorophenyl)acetic acid 97%
(2-Chloro-5-fluorophenyl)acetic acid

Precaución: for Research Use Only

Formula: (C₆H₃ClF)CH₂COOH
MDL Number: MFCD01016155
CAS Number: 177985-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR