Diethylene glycol monomethyl ether 98% stabilised
Supplier: Ambeed
|
Warning
|
Synonyms:
2-(2-Methoxyethoxy)ethanol, Diethylene glycol methyl ether
Stabilization:
Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
A124404-1000G
A124404-100G
A124404-25G
A124404-500G
77234-268EA
19
USD
77234-268
77234-270
77234-272
77234-274
Diethylene glycol monomethyl ether 98% stabilised
Diethylene glycol monomethyl ether
Precaución: for Research Use Only
Formula:
C₅H₁₂O₃ MW: 120.15 g/mol Boiling Pt: 193 Melting Pt: -65 Flash Pt: 91 |
MDL Number:
MFCD00002871 CAS Number: 111-77-3 Merck Index: 12,06116 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...