2,2-Bis(hydroxymethyl)propionic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3-Hydroxy-2-(hydroxymethyl)-2-methylpropanoic acid

A124335-1000G A124335-100G A124335-25G A124335-500G
77246-240EA 23 USD
77246-240 77246-242 77246-244 77246-246
2,2-Bis(hydroxymethyl)propionic acid 98%
2,2-Bis(hydroxymethyl)propionic acid

Precaución: for Research Use Only

Formula: C₅H₁₀O₄
MDL Number: MFCD00004199
CAS Number: 4767-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR