(4-Bromo-2,6-difluorophenyl)methanamine 97%

Supplier: Ambeed
Warning

A124171-100MG A124171-1G A124171-250MG A124171-5G
77258-172EA 40.17 USD
77258-172 77258-174 77258-176 77258-178
(4-Bromo-2,6-difluorophenyl)methanamine 97%
(4-Bromo-2,6-difluorophenyl)methanamine

Precaución: for Research Use Only

Formula: C₇H₆BrF₂N
MW: 222.03 g/mol
MDL Number: MFCD04972477
CAS Number: 887585-99-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR