Ethyl 5-bromo-2-(2,2,2-trifluoroacetamido)thiophene-3-carboxylate 97%

Supplier: Ambeed
Warning

A124117-100MG A124117-1G A124117-250MG A124117-5G
77246-220EA 22 USD
77246-220 77246-222 77246-224 77246-226
Ethyl 5-bromo-2-(2,2,2-trifluoroacetamido)thiophene-3-carboxylate 97%
Ethyl 5-bromo-2-(2,2,2-trifluoroacetamido)thiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrF₃NO₃S
MDL Number: MFCD30536240
CAS Number: 2044706-51-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR