(4-Bromothieno[2,3-c]pyridin-2-yl)methanol 97%

Supplier: Ambeed
Warning

A124039-100MG A124039-1G A124039-250MG
77234-262EA 8.24 USD
77234-262 77234-264 77234-266
(4-Bromothieno[2,3-c]pyridin-2-yl)methanol 97%
(4-Bromothieno[2,3-c]pyridin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₈H₆BrNOS
MW: 244.11 g/mol
MDL Number: MFCD12923087
CAS Number: 870243-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR