N,N-Diethyl-4-fluoro-3-nitrobenzamide 97%

Supplier: Ambeed
Warning

A123968-100MG A123968-250MG
77246-214EA 21.63 USD
77246-214 77246-216
N,N-Diethyl-4-fluoro-3-nitrobenzamide 97%
N,N-Diethyl-4-fluoro-3-nitrobenzamide

Precaución: for Research Use Only

Formula: C₁₁H₁₃FN₂O₃
MW: 240.23 g/mol
MDL Number: MFCD12408749
CAS Number: 474018-94-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR