Methyl-5-amino-2-methylbenzoate 97%
Supplier: Ambeed
|
Warning
|
A123959-100G
A123959-25G
A123959-5G
77246-206EA
83
USD
77246-206
77246-208
77246-210
77246-212
Methyl-5-amino-2-methylbenzoate 97%
Methyl-5-amino-2-methylbenzoate
Precaución: for Research Use Only
Formula:
C₉H₁₁NO₂ |
MDL Number:
MFCD08752568 CAS Number: 18595-12-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...