2-(Chloromethyl)-6-fluoropyridine 97%

Supplier: Ambeed
Danger

A123935-1G A123935-250MG
77258-168EA 459.38 USD
77258-168 77258-170
2-(Chloromethyl)-6-fluoropyridine 97%
2-(Chloromethyl)-6-fluoropyridine

Precaución: for Research Use Only

Formula: C₆H₅ClFN
MW: 145.56 g/mol
MDL Number: MFCD07774124
CAS Number: 315180-16-6
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR