tert-Butyl 2-(4-bromo-1H-pyrazol-1-yl)acetate 97%

Supplier: Ambeed
Warning

A123910-1G A123910-250MG A123910-25G A123910-5G
77234-254EA 20 USD
77234-254 77234-256 77234-258 77234-260
tert-Butyl 2-(4-bromo-1H-pyrazol-1-yl)acetate 97%
tert-Butyl 2-(4-bromo-1H-pyrazol-1-yl)acetate

Precaución: for Research Use Only

Formula: C₉H₁₃BrN₂O₂
MDL Number: MFCD13193178
CAS Number: 1199773-67-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR