N-Carbobenzyloxy-L-valine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-Cbz-L-valine, Z-L-Val-OH, Z-Val-OH
A123881-1000G
A123881-100G
A123881-10G
A123881-25G
A123881-500G
A123881-5G
77271-692EA
151.41
USD
77271-692
77271-694
77271-696
77271-698
77271-700
77271-702
N-Carbobenzyloxy-L-valine 98%
N-Carbobenzyloxy-L-valine
Precaución: for Research Use Only
Formula:
C₁₃H₁₇NO₄ MW: 251.3 g/mol |
MDL Number:
MFCD00008922 CAS Number: 1149-26-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...